Learn More
Genscript Corporation CHO-K1/FcRn Stable Cell Line
Catalog No. 5000057329
Encompass
Recombinant CHO-K1 cells stably overexpress human FcRn and B2M subunit on the surface The B2M subunits enable stable expression of FcRn in cells Association of FcRn alpha-chain with beta (2) m is also important for IgG binding The surface expression of FcRn is validated by FACS analysis
Catalog No. 50-000-57329 Supplier Genscript Corporation Supplier No. M00603
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human FcRn and B2M subunit on the surface The B2M subunits enable stable expression of FcRn in cells Association of FcRn alpha-chain with beta (2) m is also important for IgG binding The surface expression of FcRn is validated by FACS analysis