Learn More
Genscript Corporation CHO-K1/FFA1/G15 Stable Cell L
Supplier:  Genscript Corporation M00273
Encompass
Recombinant CHO-K1 cells stably overexpress human free fatty acid receptor 1 (FFA1) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58626
Add to Cart
Request a Quote