Learn More
Genscript Corporation CHO-K1/FFA2/G15 Stable Cell L
Catalog No. 5000057302
Encompass
FFA2 (GPR43) is a 330-amino acid G-protein coupled receptor that binds to fatty acid FFA2 is highly selective expressed in leukocytes which suggests a role in the recruitment of these cell populations toward sites of bacterial infection
Catalog No. 50-000-57302 Supplier Genscript Corporation Supplier No. M00437
Add to Cart
Add to Cart
Request a Quote
FFA2 (GPR43) is a 330-amino acid G-protein coupled receptor that binds to fatty acid FFA2 is highly selective expressed in leukocytes which suggests a role in the recruitment of these cell populations toward sites of bacterial infection