Learn More
Genscript Corporation CHO-K1/FPR1/G15 Stable Cell L
Catalog No. 5000058050
Encompass
The formyl peptide receptors (FPR) are a class of G protein-coupled receptors involved in chemotaxis FPR1 is a Gi-coupled GPCR FPR1 is expressed in the vasculature secretory epithelial cells neurons and other tissues FPR is involved in host defense against bacterial infection and in the clearance of damaged cells
Catalog No. 50-000-58050 Supplier Genscript Corporation Supplier No. M00429
Add to Cart
Add to Cart
Request a Quote
The formyl peptide receptors (FPR) are a class of G protein-coupled receptors involved in chemotaxis FPR1 is a Gi-coupled GPCR FPR1 is expressed in the vasculature secretory epithelial cells neurons and other tissues FPR is involved in host defense against bacterial infection and in the clearance of damaged cells