Learn More
Genscript Corporation CHO-K1/G15/AMY1 Stable Cell L
Catalog No. 5000057465
Encompass
Recombinant CHO-K1 cells stably overexpress human receptor activity modifying protein 1 (RAMP1) and calcitonin receptor (CTR) on the surface Amylin receptors (AMYR) are a class of special protein heterodimers composed of a CTR combined with one of three homologous receptor activity-modifying proteins (RAMP1 RAMP2 RAMP3) forming three subtypes (AMY1 AMY2 AMY3) This cell contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57465 Supplier Genscript Corporation Supplier No. M00557
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human receptor activity modifying protein 1 (RAMP1) and calcitonin receptor (CTR) on the surface Amylin receptors (AMYR) are a class of special protein heterodimers composed of a CTR combined with one of three homologous receptor activity-modifying proteins (RAMP1 RAMP2 RAMP3) forming three subtypes (AMY1 AMY2 AMY3) This cell contain high levels of G protein G s to couple with the receptor in downstream signaling pathways