Learn More
Genscript Corporation CHO-K1/G15/CCR5
Catalog No. 5000057304
Encompass
C-C chemokine receptor type 5 (CCR5) is a G protein-coupled receptor and a co-receptor for the entry of human immunodeficiency virus-1 (HIV-1) into cells CCR5 chemokine receptor is involved in leucocytes chemotaxis to sites of inflammation and plays an important role in the macrophages T cells and monocytes recruitment The chemokine ligands that bind to CCR5 are regulated on activation normal T Cell expressed and secreted (RANTES) and macrophage inflammatory protein 1 alpha (MIP1 ) GenScript s human CCR5-expressing stable subline is guaranteed to function properly in the calcium flux assay
Catalog No. 50-000-57304 Supplier Genscript Corporation Supplier No. M00523
Add to Cart
Add to Cart
Request a Quote
C-C chemokine receptor type 5 (CCR5) is a G protein-coupled receptor and a co-receptor for the entry of human immunodeficiency virus-1 (HIV-1) into cells CCR5 chemokine receptor is involved in leucocytes chemotaxis to sites of inflammation and plays an important role in the macrophages T cells and monocytes recruitment The chemokine ligands that bind to CCR5 are regulated on activation normal T Cell expressed and secreted (RANTES) and macrophage inflammatory protein 1 alpha (MIP1 ) GenScript s human CCR5-expressing stable subline is guaranteed to function properly in the calcium flux assay