Learn More
Genscript Corporation CHO-K1/G15/CXCR1
Catalog No. 5000057463
Encompass
Chemokine (C-X-C motif) receptor 1 (CXCR1) is a rhodopsin-like G protein-coupled receptor It is one of two high-affinity receptors for the CXC chemokine interleukin-8 (IL-8) a major mediator of immune and inflammatory responses The structure of human CXCR1 in a lipid bilayer should help to facilitate the discovery of new compounds that interact with GPCRs GenScript s human CXCR1-expressing stable subline is guaranteed to function properly in the calcium flux assay
Catalog No. 50-000-57463 Supplier Genscript Corporation Supplier No. M00524
Add to Cart
Add to Cart
Request a Quote
Chemokine (C-X-C motif) receptor 1 (CXCR1) is a rhodopsin-like G protein-coupled receptor It is one of two high-affinity receptors for the CXC chemokine interleukin-8 (IL-8) a major mediator of immune and inflammatory responses The structure of human CXCR1 in a lipid bilayer should help to facilitate the discovery of new compounds that interact with GPCRs GenScript s human CXCR1-expressing stable subline is guaranteed to function properly in the calcium flux assay