Learn More
Genscript Corporation CHO-K1/G15/CXCR3
Catalog No. 5000058656
Encompass
Chemokine (C-X-C motif) receptor 3 (CXCR3) belongs to the CXC chemokine receptor family Similar to other chemokine receptors it is a G protein-coupled receptor (GPCR) CXCR3 has two distinct splice variants CXCR3A and CXCR3B demonstrating different binding affinities CXCR3-A binds to the CXC chemokines CXCL9 (MIG) CXCL10 (IP-10) and CXCL11 (ITAC) while CXCR3-B can also bind to CXCL4 in addition to CXCL9 CXCL10 and CXCL11 GenScript s human CXCR3A-expressing stable subline is guaranteed to function properly in calcium flux assay
Catalog No. 50-000-58656 Supplier Genscript Corporation Supplier No. M00525
Add to Cart
Add to Cart
Request a Quote
Chemokine (C-X-C motif) receptor 3 (CXCR3) belongs to the CXC chemokine receptor family Similar to other chemokine receptors it is a G protein-coupled receptor (GPCR) CXCR3 has two distinct splice variants CXCR3A and CXCR3B demonstrating different binding affinities CXCR3-A binds to the CXC chemokines CXCL9 (MIG) CXCL10 (IP-10) and CXCL11 (ITAC) while CXCR3-B can also bind to CXCL4 in addition to CXCL9 CXCL10 and CXCL11 GenScript s human CXCR3A-expressing stable subline is guaranteed to function properly in calcium flux assay