Learn More
Genscript Corporation CHO-K1/G15/CXCR5
Catalog No. 5000059198
Encompass
Recombinant CHO-K1 cells stably overexpress human C-X-C motif chemokine receptor 5 (CXCR5) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-59198 Supplier Genscript Corporation Supplier No. M00526
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human C-X-C motif chemokine receptor 5 (CXCR5) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways