Learn More
Genscript Corporation CHO-K1/G15 Stable Cell Line
Catalog No. 5000058625
Encompass
Recombinant CHO-K1 cells stably overexpress human G protein subunit alpha 15 (G 15) in the cytoplasm This protein enables G protein-coupled receptor binding activity and acts upstream of or within with a positive effect on calcium-mediated signaling
Catalog No. 50-000-58625 Supplier Genscript Corporation Supplier No. M00257
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human G protein subunit alpha 15 (G 15) in the cytoplasm This protein enables G protein-coupled receptor binding activity and acts upstream of or within with a positive effect on calcium-mediated signaling