Learn More
Genscript Corporation CHO-K1/GALR1/G15 Stable Cell
Catalog No. 5000058267
Encompass
The neuropeptide galanin elicits a range of biological effects by interacting with specific G-protein-coupled receptors Galanin receptors are seven-transmembrane proteins that have been shown to activate a variety of intracellular second-messenger pathways GALR1 inhibits adenylyl cyclase via a G protein of the Gi/Go family GALR1 is widely expressed in the brain spinal cord and peripheral sites including small intestine and heart
Catalog No. 50-000-58267 Supplier Genscript Corporation Supplier No. M00277
Add to Cart
Add to Cart
Request a Quote
The neuropeptide galanin elicits a range of biological effects by interacting with specific G-protein-coupled receptors Galanin receptors are seven-transmembrane proteins that have been shown to activate a variety of intracellular second-messenger pathways GALR1 inhibits adenylyl cyclase via a G protein of the Gi/Go family GALR1 is widely expressed in the brain spinal cord and peripheral sites including small intestine and heart