Learn More
Genscript Corporation CHO-K1/GALR2 Stable Cell Line
Catalog No. 5000057077
Encompass
Recombinant CHO-K1 cells stably overexpress human galanin receptor 2 (GALR2) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57077 Supplier Genscript Corporation Supplier No. M00209
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human galanin receptor 2 (GALR2) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways