Learn More
Genscript Corporation CHO-K1/GCGR/G15 Stable Cell L
Catalog No. 5000057300
Encompass
Recombinant CHO-K1 cells stably overexpress human glucagon receptor (GCGR) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57300 Supplier Genscript Corporation Supplier No. M00345
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human glucagon receptor (GCGR) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways