Learn More
Genscript Corporation CHO-K1/GLP1R/G15 Stable Cell
Catalog No. 5000057303
Encompass
Recombinant CHO-K1 cells stably overexpress human glucagon-like peptide-1 receptor (GLP1R) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57303 Supplier Genscript Corporation Supplier No. M00451
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human glucagon-like peptide-1 receptor (GLP1R) on the surface and contain high levels of G protein G s to couple with the receptor in downstream signaling pathways