Learn More
Genscript Corporation CHO-K1/GLP2R/G15 Stable Cell
Catalog No. 5000058268
Encompass
The glucagon-like peptides include glucagon GLP-1 and GLP-2 exert diverse actions on nutrient intake gastrointestinal motility islet hormone secretion cell proliferation and apoptosis as well as nutrient absorptionand assimilation Glucagon-like peptide (GLP)-2 a product of the proglucagon gene is expressed in enteroendocrine cells of the small and large intestine and is trophic to the gastrointestinal mucosa GLP-2 also inhibits gastric acid secretion and emptying and up-regulates intestinal hexose transport GLP-2 acts via binding to a single G protein-coupled GLP-2 receptor (GLP-2R)
Catalog No. 50-000-58268 Supplier Genscript Corporation Supplier No. M00307
Add to Cart
Add to Cart
Request a Quote
The glucagon-like peptides include glucagon GLP-1 and GLP-2 exert diverse actions on nutrient intake gastrointestinal motility islet hormone secretion cell proliferation and apoptosis as well as nutrient absorptionand assimilation Glucagon-like peptide (GLP)-2 a product of the proglucagon gene is expressed in enteroendocrine cells of the small and large intestine and is trophic to the gastrointestinal mucosa GLP-2 also inhibits gastric acid secretion and emptying and up-regulates intestinal hexose transport GLP-2 acts via binding to a single G protein-coupled GLP-2 receptor (GLP-2R)