Learn More
Genscript Corporation CHO-K1/GPBAR1 Stable Cell Line
Catalog No. 5000058734
Encompass
The gene GPBRA1 encodes G protein-coupled bile acid receptor Bile acid-binding induces its internalization activation of extracellular signal-regulated kinase and intracellular cAMP production
Catalog No. 50-000-58734 Supplier Genscript Corporation Supplier No. M00432
Add to Cart
Add to Cart
Request a Quote
The gene GPBRA1 encodes G protein-coupled bile acid receptor Bile acid-binding induces its internalization activation of extracellular signal-regulated kinase and intracellular cAMP production