Learn More
Genscript Corporation CHO-K1/GPR68/G15 Stable Cell
Catalog No. 5000058495
Encompass
Recombinant CHO-K1 cells stably overexpress human G protein-coupled receptor 68 (GPR68) on the surface and contain high levels of G protein G i and G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58495 Supplier Genscript Corporation Supplier No. M00439
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human G protein-coupled receptor 68 (GPR68) on the surface and contain high levels of G protein G i and G q to couple with the receptor in downstream signaling pathways