Learn More
Genscript Corporation CHO-K1/H2/G15 Stable Cell Lin
Supplier:  Genscript Corporation M00306
Encompass
Histamine is a ubiquitous messenger molecule released from mast cells enterochromaffin-like cells and neurons Its various actions are mediated by histamine receptors H1 H2 H3 and H4 Specifically the histamine receptor H2 belongs to the rhodopsin-like family of G protein-coupled receptors As an integral membrane protein it stimulates gastric acid secretion and regulates gastrointestinal motility and intestinal secretion and is thought to be involved in regulating cell growth and differentiation
Catalog No. 50-000-59121
Add to Cart
Request a Quote