Learn More
Genscript Corporation CHO-K1/H3/G15 Stable Cell Lin
Catalog No. 5000059049
Encompass
The histamine receptor H3 is a G sub i /sub -coupled GPCR expressed in the thalamus caudate nucleus putamen cerebellum amygdala substantia nigra hippocampus hypothalamus and cerebral cortex Activation of H3 receptor in human nasal mucosa inhibits sympathetic vasoconstriction Expression of human H3 receptor splice variants in CHO cells shows that a deletion in the 2nd transmembrane domain alters ligand binding and deletions in the 3rd intracellular loop result in signal transduction being abolished
Catalog No. 50-000-59049 Supplier Genscript Corporation Supplier No. M00331
Add to Cart
Add to Cart
Request a Quote
The histamine receptor H3 is a G sub i /sub -coupled GPCR expressed in the thalamus caudate nucleus putamen cerebellum amygdala substantia nigra hippocampus hypothalamus and cerebral cortex Activation of H3 receptor in human nasal mucosa inhibits sympathetic vasoconstriction Expression of human H3 receptor splice variants in CHO cells shows that a deletion in the 2nd transmembrane domain alters ligand binding and deletions in the 3rd intracellular loop result in signal transduction being abolished