Learn More
Genscript Corporation CHO-K1/H4/G15 Stable Cell Lin
Supplier:  Genscript Corporation M00289
Encompass
Recombinant CHO-K1 cells stably overexpress human histamine receptor H4 (H4) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-59119
Add to Cart
Request a Quote