Learn More
Genscript Corporation CHO-K1/human CD32A 131His Stab
Catalog No. 5000058503
Encompass
Recombinant CHO-K1 cells stably overexpress human immunoglobulin gamma Fc region receptor II-a (Fc RIIa/CD32A 131His) on the surface The surface expression of CD32A 131His is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies and Fc fusion proteins or antibody based biologics binding with CD32A 131His GenScript also offers CD32A 131Arg stable cell line (Cat No M00887) for Fc RIIa polymorphism study
Catalog No. 50-000-58503 Supplier Genscript Corporation Supplier No. M00598
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human immunoglobulin gamma Fc region receptor II-a (Fc RIIa/CD32A 131His) on the surface The surface expression of CD32A 131His is validated by FACS analysis This stable cell line product is designed for measuring binding affinity and stability of Fc region of antibodies and Fc fusion proteins or antibody based biologics binding with CD32A 131His GenScript also offers CD32A 131Arg stable cell line (Cat No M00887) for Fc RIIa polymorphism study