Learn More
Genscript Corporation CHO-K1/human CD47 Stable Cell
Catalog No. 5000058758
Encompass
Recombinant CHO-K1 cells stably overexpress human CD47 on the surface The surface expression of CD47 on this cell is validated by FACS analysis This cell line is recommended for cell-based binding assay to screening antibodies against CD47 or to measure binding affinity between CD47 and anti-CD47 antibodies
Catalog No. 50-000-58758 Supplier Genscript Corporation Supplier No. M00581
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human CD47 on the surface The surface expression of CD47 on this cell is validated by FACS analysis This cell line is recommended for cell-based binding assay to screening antibodies against CD47 or to measure binding affinity between CD47 and anti-CD47 antibodies