Learn More
Genscript Corporation CHO-K1/ICOS Stable Cell Line
Catalog No. 5000057330
Encompass
The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family ICOS (Inducible T-cell COStimulator) is expressed on activated T cells It forms homodimers and plays an important role in cell-cell signaling immune responses and regulation of cell proliferation It is thought to be important for Th2 cells in particular
Catalog No. 50-000-57330 Supplier Genscript Corporation Supplier No. M00649
Add to Cart
Add to Cart
Request a Quote
The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family ICOS (Inducible T-cell COStimulator) is expressed on activated T cells It forms homodimers and plays an important role in cell-cell signaling immune responses and regulation of cell proliferation It is thought to be important for Th2 cells in particular