Learn More
Genscript Corporation CHO-K1/ITGAE Stable Cell Line
Catalog No. 5000058072
Encompass
Integrin alpha E (ITGAE) also known as CD103 (cluster of differentiation 103) is an integrin protein that in human ITGAE is expressed widely on intraepithelial lymphocyte (IEL) T cells (both T cells and T cells) and on some peripheral regulatory T cells (Tregs) It has also been reported on lamina propria T cells A subset of dendritic cells in the gut mucosa and mesenteric lymph nodes known as CD103 dendritic cells also expresses this marker
Catalog No. 50-000-58072 Supplier Genscript Corporation Supplier No. M00592
Add to Cart
Add to Cart
Request a Quote
Integrin alpha E (ITGAE) also known as CD103 (cluster of differentiation 103) is an integrin protein that in human ITGAE is expressed widely on intraepithelial lymphocyte (IEL) T cells (both T cells and T cells) and on some peripheral regulatory T cells (Tregs) It has also been reported on lamina propria T cells A subset of dendritic cells in the gut mucosa and mesenteric lymph nodes known as CD103 dendritic cells also expresses this marker