Learn More
Genscript Corporation CHO-K1/KIR Stable Cell Line
Catalog No. 5000058742
Encompass
Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and are subsets of T cells The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM) while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals The ligands for several KIR proteins are subsets of HLA class I molecules thus KIR proteins are thought to play an important role in regulation of the immune response
Catalog No. 50-000-58742 Supplier Genscript Corporation Supplier No. M00650
Add to Cart
Add to Cart
Request a Quote
Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and are subsets of T cells The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM) while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals The ligands for several KIR proteins are subsets of HLA class I molecules thus KIR proteins are thought to play an important role in regulation of the immune response