Learn More
Genscript Corporation CHO-K1/KiSS1 Stable Cell Line
Catalog No. 5000058261
Encompass
Recombinant CHO-K1 cells stably overexpress human KiSS1 receptor (KiSS1R) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58261 Supplier Genscript Corporation Supplier No. M00190
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human KiSS1 receptor (KiSS1R) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways