Learn More
Genscript Corporation CHO-K1/M3 Stable Cell Line
Catalog No. 5000057389
Encompass
Recombinant CHO-K1 cells stably overexpress human cholinergic receptor muscarinic 3 (M3) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57389 Supplier Genscript Corporation Supplier No. M00259
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human cholinergic receptor muscarinic 3 (M3) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways