Learn More
Genscript Corporation CHO-K1/M4/G15 Stable Cell Lin
Catalog No. 5000058041
Encompass
Recombinant CHO-K1 cells stably overexpress human cholinergic receptor muscarinic 4 (M4) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58041 Supplier Genscript Corporation Supplier No. M00238
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human cholinergic receptor muscarinic 4 (M4) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways