Learn More
Genscript Corporation CHO-K1/M5 Stable Cell Line
Catalog No. 5000058378
Encompass
Muscarinic acetylcholine receptors belong to a superfamily of seven-TM-domain receptors that interact with G-proteins to initiate intracellular responses Five muscarinic receptor subtypes have been identified named M1 through M5 Receptors of the M5 receptor subtype couple through the G sub q/11 /sub class of G-proteins and activate the phospholipase C pathway Activation of this pathway in turn leads to increases in free intracellular calcium levels as inositol triphosphate mediates release of calcium from the endoplasmic reticulum RT-PCR reveals that M5 mRNA is quite uniformly expressed in brain However there is little data regarding the expression and function of the M5 receptor in peripheral tissues Currently it is clear that the M5 receptor due to the high likelihood that its distribution is restricted to the CNS probably plays a discrete role in dopaminergic transmission Although the identification of M5 expression in salivary glands and iris-ciliary muscle sugge
Catalog No. 50-000-58378 Supplier Genscript Corporation Supplier No. M00186
Add to Cart
Add to Cart
Request a Quote
Muscarinic acetylcholine receptors belong to a superfamily of seven-TM-domain receptors that interact with G-proteins to initiate intracellular responses Five muscarinic receptor subtypes have been identified named M1 through M5 Receptors of the M5 receptor subtype couple through the G sub q/11 /sub class of G-proteins and activate the phospholipase C pathway Activation of this pathway in turn leads to increases in free intracellular calcium levels as inositol triphosphate mediates release of calcium from the endoplasmic reticulum RT-PCR reveals that M5 mRNA is quite uniformly expressed in brain However there is little data regarding the expression and function of the M5 receptor in peripheral tissues Currently it is clear that the M5 receptor due to the high likelihood that its distribution is restricted to the CNS probably plays a discrete role in dopaminergic transmission Although the identification of M5 expression in salivary glands and iris-ciliary muscle sugge