Learn More
Genscript Corporation CHO-K1/MC2/G15 Stable Cell Li
Catalog No. 5000057856
Encompass
The melanocortin receptor 2 MC2 receptor is G sub s /sub -coupled GPCRs expressed in zona fasciculata of the adrenal cortex placental and stimulates production of cortisol MC2 is a member of the rhodopsin family of 7-transmembrane and it s also known as the ACTH receptor or corticotropin receptor because it is specific for ACTH alone Activation of the MC2 receptor initiates a cascade of events affecting multiple steps in corticoid steroidogenesis Mutations in MC2 may result in familial glucocorticoid deficiency a group of autosomal recessive disorders characterized by resistance to ACTH
Catalog No. 50-000-57856 Supplier Genscript Corporation Supplier No. M00336
Add to Cart
Add to Cart
Request a Quote
The melanocortin receptor 2 MC2 receptor is G sub s /sub -coupled GPCRs expressed in zona fasciculata of the adrenal cortex placental and stimulates production of cortisol MC2 is a member of the rhodopsin family of 7-transmembrane and it s also known as the ACTH receptor or corticotropin receptor because it is specific for ACTH alone Activation of the MC2 receptor initiates a cascade of events affecting multiple steps in corticoid steroidogenesis Mutations in MC2 may result in familial glucocorticoid deficiency a group of autosomal recessive disorders characterized by resistance to ACTH