Learn More
Genscript Corporation CHO-K1/MC3/G15 Stable Cell Li
Catalog No. 5000057084
Encompass
The melanocortin receptor 3 MC3 receptor is Gs-coupled GPCRs expressed in brain placental and gut tissues but not in melanoma cells or in the adrenal gland MC3 receptor knockouts exhibit a metabolic syndrome At 4-6 months old MC3 receptor knockout mice show increased fat mass reduced lean mass and a higher feed efficiency with normal metabolic rates MC3 receptor knockout mice are hyperleptinaemic and males are often mildly hyperinsulinaemic
Catalog No. 50-000-57084 Supplier Genscript Corporation Supplier No. M00242
Add to Cart
Add to Cart
Request a Quote
The melanocortin receptor 3 MC3 receptor is Gs-coupled GPCRs expressed in brain placental and gut tissues but not in melanoma cells or in the adrenal gland MC3 receptor knockouts exhibit a metabolic syndrome At 4-6 months old MC3 receptor knockout mice show increased fat mass reduced lean mass and a higher feed efficiency with normal metabolic rates MC3 receptor knockout mice are hyperleptinaemic and males are often mildly hyperinsulinaemic