Learn More
Genscript Corporation CHO-K1/MC4/G15 cAMP Stable
Catalog No. 5000056739
Encompass
Human Recombinant CHO-K1/MC4/G 15 (cAMP) Stable Cell Line stably overexpress the melanocortin 4 receptor gene (MC4) This stable cell line product is designed for measuring the In Vitro function of MC4
Catalog No. 50-000-56739 Supplier Genscript Corporation Supplier No. M01141
Add to Cart
Add to Cart
Request a Quote
Human Recombinant CHO-K1/MC4/G 15 (cAMP) Stable Cell Line stably overexpress the melanocortin 4 receptor gene (MC4) This stable cell line product is designed for measuring the In Vitro function of MC4