Learn More
Genscript Corporation CHO-K1/MC5/G15 Stable Cell Li
Catalog No. 5000058265
Encompass
The melanocortin receptor 5 MC5 receptor is Gs-coupled GPCRs expressed in the adrenal gland fat cells kidney leukocytes lung lymph node mammary gland ovary pituitary testis and uterus Mice with targeted deletion of the MC5 receptor have a defect in water and thermoregulation secondary to their decreased production of sebaceous lipids
Catalog No. 50-000-58265 Supplier Genscript Corporation Supplier No. M00243
Add to Cart
Add to Cart
Request a Quote
The melanocortin receptor 5 MC5 receptor is Gs-coupled GPCRs expressed in the adrenal gland fat cells kidney leukocytes lung lymph node mammary gland ovary pituitary testis and uterus Mice with targeted deletion of the MC5 receptor have a defect in water and thermoregulation secondary to their decreased production of sebaceous lipids