Learn More
Genscript Corporation CHO-K1/MCH2 Stable Cell Line
Catalog No. 5000057390
Encompass
Coupled exclusively Gq MCH2 is a receptor for melanin-concentrating hormone Other than regulating skin color two subtypes (MCHR1 and MCHR2) are involved in energy homeostasis and social behaviors In contrast to MCHR1 that is conserved among all mammals functional MCHR2 is only expressed in carnivores and primates but not in rodents
Catalog No. 50-000-57390 Supplier Genscript Corporation Supplier No. M00280
Add to Cart
Add to Cart
Request a Quote
Coupled exclusively Gq MCH2 is a receptor for melanin-concentrating hormone Other than regulating skin color two subtypes (MCHR1 and MCHR2) are involved in energy homeostasis and social behaviors In contrast to MCHR1 that is conserved among all mammals functional MCHR2 is only expressed in carnivores and primates but not in rodents