Learn More
Genscript Corporation CHO-K1/mouse ADORA2A Stable Ce
Supplier:  Genscript Corporation M00575
Encompass
The adenosine receptors ADORA2A are Gs-coupled GPCRs expressed in the thymus gland heart lung kidney brain platelets spleen and leukocytes ADORA2A down-regulates chemokine receptor function and inhibits platelet aggregation ADORA2A antagonists may be useful as therapy for Parkinson s disease
Catalog No. 50-000-58757
Add to Cart
Request a Quote