Learn More
Genscript Corporation CHO-K1/Mouse CD47 Stable Cell
Supplier: Genscript Corporation M00631
This gene encodes a membrane protein which is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix The encoded protein is also a receptor for the C-terminal cell binding domain of thrombospondin and it may play a role in membrane transport and signal transduction This gene has broad tissue distribution and is reduced in expression on Rh erythrocytes Alternatively spliced transcript variants have been found for this gene