Learn More
Genscript Corporation CHO-K1/mouse CTLA4 Stable Cell
Catalog No. 5000058524
Encompass
Recombinant CHO-K1 cells stably overexpress Mus musculus cytotoxic T-lymphocyte-associated protein 4 (CTLA4) on the surface The surface expression of mouse CTLA4 is validated by FACS analysis This cell line is designed for cell-based binding for screening antibodies binding with mouse CTLA4 and for evaluating target binding affinity
Catalog No. 50-000-58524 Supplier Genscript Corporation Supplier No. M00570
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress Mus musculus cytotoxic T-lymphocyte-associated protein 4 (CTLA4) on the surface The surface expression of mouse CTLA4 is validated by FACS analysis This cell line is designed for cell-based binding for screening antibodies binding with mouse CTLA4 and for evaluating target binding affinity