Learn More
Genscript Corporation CHO-K1/mouse HVEM Stable Cell
Supplier:  Genscript Corporation M00647
Encompass
HVEM is found on the surface of various cell types including hematopoietic and non-hematopoietic cells and is expressed mainly in spleen thymus bone marrow lung and intestines It has been identified as a canonical TNF receptor signaling through the TNF receptor-associated factors (TRAFs) leading to NF B activation Furthermore in addition to acting as a signaling receptor in binding to Ig superfamily members it acts as a ligand for these inhibitory receptors Therefore bidirectional signaling is possible for the HVEM-mediated signaling network which can be involved in positive or negative immunological reactions under different contexts
Catalog No. 50-000-58679
Only null left
Add to Cart
Request a Quote