Learn More
Genscript Corporation CHO-K1/mouse PD-L1 Stable Cell
Catalog No. 5000058523
Encompass
Recombinant CHO-K1 cells stably overexpress mouse programmed cell death 1 ligand 1 (PD-L1) on the surface The surface expression of PD-L1 is validated by FACS analysis
Catalog No. 50-000-58523 Supplier Genscript Corporation Supplier No. M00567
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress mouse programmed cell death 1 ligand 1 (PD-L1) on the surface The surface expression of PD-L1 is validated by FACS analysis