Learn More
Genscript Corporation CHO-K1/mouse TIGIT Stable Cell
Supplier:  Genscript Corporation M00585
Encompass
TIGIT (also called T cell immunoreceptor with Ig and ITIM domains) is one newly discovered immune receptor on some percentage of T cells and Natural Killer Cells (NK) It is also identified as WUCAM and Vstm3 TIGIT binds to CD155 (PVR) on dendritic cells (DCs) macrophages etc with high affinity and also to CD112 (PVRL2) with lower affinity Research has shown that TIGIT-Fc fusion proteins interact with PVR on dendritic cells and increase IL-10 secretion while decreasing IL-12 secretion under LPS stimulation TIGIT-Fc fusion proteins also inhibit T cell activation in vivo TIGIT s inhibition of NK cytotoxicity can be blocked by antibodies against its interaction with PVR which is directed through its ITIM domain
Catalog No. 50-000-59249
Add to Cart
Request a Quote