Learn More
Genscript Corporation CHO-K1/Mouse VISTA Stable Cell
Catalog No. 5000058073
Encompass
V-domain Ig suppressor of T cell activation (VISTA) is a potent negative regulator of T-cell function that is expressed on hematopoietic cells VISTA levels are heightened within the tumor microenvironment in which its blockade can enhance antitumor immune responses in mice In humans blockade of the related programmed cell death 1 (PD-1) pathway has shown great potential in clinical immunotherapy trials
Catalog No. 50-000-58073 Supplier Genscript Corporation Supplier No. M00633
Add to Cart
Add to Cart
Request a Quote
V-domain Ig suppressor of T cell activation (VISTA) is a potent negative regulator of T-cell function that is expressed on hematopoietic cells VISTA levels are heightened within the tumor microenvironment in which its blockade can enhance antitumor immune responses in mice In humans blockade of the related programmed cell death 1 (PD-1) pathway has shown great potential in clinical immunotherapy trials