Learn More
Genscript Corporation CHO-K1/MRGPRX2 Stable Cell Lin
Supplier:  Genscript Corporation M00425
Encompass
Recombinant CHO-K1 cells stably overexpress human mas-related G-protein coupled receptor member X2 (MRGPRX2) on the surface and contain high levels of G protein G q and G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-56747
Add to Cart
Request a Quote