Learn More
Genscript Corporation CHO-K1/MT2/G15 Stable Cell Li
Catalog No. 5000057097
Encompass
Recombinant CHO-K1 cells stably overexpress human melatonin receptor 1B (MT2) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57097 Supplier Genscript Corporation Supplier No. M00312
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human melatonin receptor 1B (MT2) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways