Learn More
Genscript Corporation CHO-K1/MTLR/G15 Stable Cell L
Catalog No. 5000057250
Encompass
The motilin receptor (MTLR) represents a clinically useful pharmacological target as agonists binding to the MTLR have gastroprokinetic properties Motilin is a 22 amino acid peptide that potently stimulates gastrointestinal contractility The biological effects of motilin are mediated by a G sub q /sub -coupled seven transmembrane protein currently termed motilin receptor (MR) that shares significant sequence similarity with the ghrelin receptor The motilin receptor is also activated by the antibiotic erythromycin this interaction appears to mediate some of the gastrointestinal side effects of erythromycin Although motilides (non-antibiotic derivatives of erythromycin) such as ABT-229 have been investigated for treatment of diabetic gastroporesis the effectiveness has been limited by tachyphylaxis (decreased response to ligand) resulting from receptor downregulation Agonists of the motilin receptor with reduced densensitization activity remain a potential treatment for dis
Catalog No. 50-000-57250 Supplier Genscript Corporation Supplier No. M00347
Add to Cart
Add to Cart
Request a Quote
The motilin receptor (MTLR) represents a clinically useful pharmacological target as agonists binding to the MTLR have gastroprokinetic properties Motilin is a 22 amino acid peptide that potently stimulates gastrointestinal contractility The biological effects of motilin are mediated by a G sub q /sub -coupled seven transmembrane protein currently termed motilin receptor (MR) that shares significant sequence similarity with the ghrelin receptor The motilin receptor is also activated by the antibiotic erythromycin this interaction appears to mediate some of the gastrointestinal side effects of erythromycin Although motilides (non-antibiotic derivatives of erythromycin) such as ABT-229 have been investigated for treatment of diabetic gastroporesis the effectiveness has been limited by tachyphylaxis (decreased response to ligand) resulting from receptor downregulation Agonists of the motilin receptor with reduced densensitization activity remain a potential treatment for dis