Learn More
Genscript Corporation CHO-K1/NIACR1/G15 Stable Cell
Catalog No. 5000057299
Encompass
Recombinant CHO-K1 cells stably overexpress human nicotinic acid receptor (NIACR1) also known as hydroxycarboxylic acid receptor 2 (HCAR2) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57299 Supplier Genscript Corporation Supplier No. M00428
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human nicotinic acid receptor (NIACR1) also known as hydroxycarboxylic acid receptor 2 (HCAR2) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways