Learn More
Genscript Corporation CHO-K1/NK1 Stable Cell Line
Catalog No. 5000057081
Encompass
Tachykinins are peptides sharing the common C-terminal amino acid sequence Phe-X-Gly-Leu-Met-NH sub 2 /sub This neuropeptide family is composed of substance P neurokinin A and neurokinin B which are widely distributed in mammalian central and peripheral nervous systems It plays a role as both a neurotransmitter and a neuromodulator Their actions are mediated by binding with three distinct receptors namely NK1 NK2 and NK3 NK1 has high affinity with substance P In the CNS NK1 has been implicated to play a role in regulating neuronal survival and degeneration In the cardiovascular system NK1 mediates endothelium-dependent vasodilatation and plasma protein extravasations In the gastrointestinal system NK1 receptors mediate intestinal motility secretion and vascular functions SP-NK1 receptor communication is also involved in glioma development and progression NK1 receptor antagonists may have several therapeutic applications in diseases mediated by tachykinins such as
Catalog No. 50-000-57081 Supplier Genscript Corporation Supplier No. M00199
Add to Cart
Add to Cart
Request a Quote
Tachykinins are peptides sharing the common C-terminal amino acid sequence Phe-X-Gly-Leu-Met-NH sub 2 /sub This neuropeptide family is composed of substance P neurokinin A and neurokinin B which are widely distributed in mammalian central and peripheral nervous systems It plays a role as both a neurotransmitter and a neuromodulator Their actions are mediated by binding with three distinct receptors namely NK1 NK2 and NK3 NK1 has high affinity with substance P In the CNS NK1 has been implicated to play a role in regulating neuronal survival and degeneration In the cardiovascular system NK1 mediates endothelium-dependent vasodilatation and plasma protein extravasations In the gastrointestinal system NK1 receptors mediate intestinal motility secretion and vascular functions SP-NK1 receptor communication is also involved in glioma development and progression NK1 receptor antagonists may have several therapeutic applications in diseases mediated by tachykinins such as