Learn More
Genscript Corporation CHO-K1/NPS1b/G15 Stable Cell
Catalog No. 5000058049
Encompass
Recombinant CHO-K1 cells stably overexpress human neuropeptide S receptor 1 (NPSR1) isoform B on the surface and contain high levels of G protein G s and G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58049 Supplier Genscript Corporation Supplier No. M00344
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human neuropeptide S receptor 1 (NPSR1) isoform B on the surface and contain high levels of G protein G s and G q to couple with the receptor in downstream signaling pathways