Learn More
Genscript Corporation CHO-K1/NPY4/G15 Stable Cell L
Catalog No. 5000059118
Encompass
Recombinant CHO-K1 cells stably overexpress human neuropeptide Y receptor Y4 (NPY4) on the surface and contain high levels of G protein G i and G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-59118 Supplier Genscript Corporation Supplier No. M00237
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human neuropeptide Y receptor Y4 (NPY4) on the surface and contain high levels of G protein G i and G q to couple with the receptor in downstream signaling pathways