Learn More
Genscript Corporation CHO-K1/OPRD1/G15 Stable Cell
Catalog No. 5000058042
Encompass
Recombinant CHO-K1 cells stably overexpress human opioid receptor delta 1 (OPRD1) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-58042 Supplier Genscript Corporation Supplier No. M00323
Add to Cart
Add to Cart
Request a Quote
Recombinant CHO-K1 cells stably overexpress human opioid receptor delta 1 (OPRD1) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways