Learn More
Genscript Corporation CHO-K1/OX-40L Stable Cell Line
Supplier:  Genscript Corporation M00563
Encompass
Recombinant CHO-K1 cells stably overexpress human OX-40L on the surface The surface expression of OX-40L is validated by FACS analysis
Catalog No. 50-000-58658
Only null left
Add to Cart
Request a Quote